Implen C40 NanoPhotometer UV/Vis Spectrophotometer with Touch Screen, 110/220 V

Representative image only.
Implen Mfr # C40-Touch – Item # EW-83070-60
Microvolume measurement with on-the-go analysis
  • Flexible control and data collection via computer, Mac, smartphone, or tablet
  • Choose a Touch model for a built-in touch screen tablet
  • Slot for a Kensington™ lock provides antitheft security
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Source LampXenon
  • Bandwidth< 1.8 nm
  • Min Wavelength (nm)200
  • Max Wavelength (nm)900
  • Wavelength Accuracy±0.75 nm
  • Wavelength Reproducibility±0.2 nm
  • Pathlength (mm)0.67 and 0.07
  • Min Photometric - Absorbance (A)0
  • Max Photometric - Absorbance (A)2.6
  • Detector3648 CCD Array
  • Concentration130 ng/µL (ds DNA); 3.7 mg/mL (BSA)
  • Scan Time3.5 to 6 seconds
  • Display TypeTouch screen
  • Width (cm)20
  • Height (cm)12
  • Depth (cm)20
  • Width (in)7 7/8
  • Height (in)4 3/4
  • Depth (in)7 7/8
  • Power (VAC)90 to 250
  • Power (Hz)50/60
  • DescriptionNanoPhotometer UV/Vis Spectrophotometer with Touch Screen, 110/220 V
  • Warranty1 year
1 Year Warranty

Key Features

  • Flexible control and data collection via computer, Mac, smartphone, or tablet
  • Choose a Touch model for a built-in touch screen tablet
  • Slot for a Kensington™ lock provides antitheft security
  • 1 year warranty

More About this Item

For those in need of flexibility, this family of UV/visible spectrophotometers offers reliable protein and DNA measurements for mobile and laboratory applications. Requiring only a minute sample size of 0.3 µL and with a limit of detection down to 0.1 ng/µL (ds DNA) or 0.003 mg/µL (BSA), this spectrophotometer series accommodates any application. Simplified optics with two precisely defined pathlengths provide two fixed and sealed anchor points, prohibiting drift and eliminating the need for recalibration.


Equipped with Wi-Fi, USB A/B, HDMI and LAN interface connections for flexible instrument control and data saving options. Front USB flash drive port offers quick and easy data transfer.


REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat