InnoCal A2LA ISO17025 Accredited Calibration; Multiparameter Timer
Representative image only.
InnoCal Item # EW-17500-56
Quick and accurate timer calibrations accredited to ISO/IEC 17025 standards
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specification EPA, USDA, ISO and GLP/GMPs standards
  • Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
PRODUCT OPTIONS
$278.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeA2LA ISO17025 Accredited Mechanical
  • System Calibration (Instrument and Probe)Yes
  • DescriptionA2LA ISO17025 Accredited Calibration; Multiparameter Timer
Key Features
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specification EPA, USDA, ISO and GLP/GMPs standards
  • Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
More About this Item
Calibrations on timers and stopwatches are performed using a precision timometer. Measurements can be taken from 60 seconds to 720 hours.