This item is now discontinued. Please browse similar items
InnoCal NIST-Traceable Calibration; Oven
Representative image only.
InnoCal Item # EW-17002-30
Ensure your instrument remains precise
  • Receive your product calibrated and ready to use
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • Order A2LA endorsement separately
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeTemperature
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points1
  • Calibration Test PointsMid-Point unless otherwise specified
  • Calibration Test Range-80 to 660C
  • DescriptionNIST-Traceable Calibration for Ovens
Key Features
  • Receive your product calibrated and ready to use
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • Order A2LA endorsement separately
More About this Item
InnoCal provides both in-house and on-site. NIST-traceable calibration services for refrigerators, freezers, incubators, and ovens. Keep your sample preparation, sample storage, or testing equipment’s temperature control system accurate and reliable. Regulatory and quality agencies like FDA and ISO have strict guidelines for the use of refrigerators, freezers, incubators, and ovens. Maintain your compliance with the help of InnoCal. Testing can be done either at a single set point or through the range depending on your requirements. Accuracy is confirmed using high-accuracy Precision Reference Thermometers (PRT’s). InnoCal’s metrologists can service most any brand of constant temperature equipment including Binder, Boekel, Carbolite, Fisher Scientific Isotemp, IKA, Lab Companion, Lab-Line, Labnet, Marvel, Nor-Lake, Precision, Quincy Lab, Salvis, ScienTemp, StableTemp, Thermo Scientific (Forma, Heratherm, MaxQ, Revco), and Yamato.