InnoCal NIST-Traceable Calibration; Stirrer and Heat (Stirring Hot Plate)
Representative image only.
InnoCal Item # EW-17002-27
Quick and accurate mixer and stirrer calibrations accredited to ISO/IEC 17025 standards
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specification EPA, USDA, ISO and GLP/GMPs standards
  • Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
PRODUCT OPTIONS
$245.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeTemperature
  • System Calibration (Instrument and Probe)Yes
  • Calibration Test PointsAcross the Range of the Instrument
  • DescriptionNIST Calibration for Stirrers and Heat
Key Features
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specification EPA, USDA, ISO and GLP/GMPs standards
  • Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
More About this Item
InnoCal provides calibration services for a wide variety of laboratory and industrial mixing and stirring equipment.
Stirrers and stirring hotplates can also be calibrated for rotational accuracy (rpm). All rotational motion (rpm) measurements are traceable to NIST and include data. Equipment is tested at 10 rpm values throughout the range. Accuracy is confirmed using NIST-traceable optical devices.
Service for stirring hotplates and hotplates adds NIST-traceable, temperature control system calibration.
Major brands of stirrers and stirring hotplates that InnoCal can service are Cole-Parmer, Corning, EchoTherm, Fisher Scientific Isotemp, IKA, Thermo Scientific (Cimarec, Super Nuova), Troemner.