InnoCal NIST-Traceable Calibration; Torque Meter/Tester
Representative image only.
InnoCal Item # EW-17100-41
NIST-traceable calibration for torque wrenches, screwdrivers, testers, and more!
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeForce and Torque
  • Instrument Only CalibrationYes
  • Number Of Calibration Test Points5 Points Clockwise, 5 Points Counter-Clockwise
  • Calibration Test PointsAcross range
  • Calibration Test Range0 to 2000 ft-lbf
  • DescriptionNIST-Traceable Calibration; Torque Meter/Tester
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005