InnoCal NIST-Traceable Calibration; Viscometer
Representative image only.
InnoCal Item # UX-17107-01
Ensure your instrument remains precise
  • Receive your product calibrated and ready to use
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • Order A2LA endorsement separately
  • Calibration service for a viscometer
PRODUCT OPTIONS
$343.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeLiquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points1 Per Spindle
  • Calibration Test PointsApproximately Mid-Point of Range
  • DescriptionNIST Calibration for Viscometers
Key Features
  • Receive your product calibrated and ready to use
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • Order A2LA endorsement separately
  • Calibration service for a viscometer
More About this Item
Rotational viscometers are tested at the midpoint value of the spindles. Accuracy is confirmed using ISO Guide 34 Accredited Standard Reference Materials.