International Enviroguard Coveralls, elastic wrists, large, 25/cs SKU 8622626
Representative image only.
International Enviroguard Mfr # 4012-L – Item # EW-86226-26
Fabric retains >95% of 0.3 micron particulates while allowing high air flow for breathability
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Protects against dangerous noxious particulates
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • ColorWhite
  • DescriptionCoveralls, elastic wrists, large, 25/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Protects against dangerous noxious particulates
More About this Item
These coveralls are constructed of electrostatically-charged filter media for superior protection against particulates. Polymer grid on the exterior surface provides exceptional fabric strength and abrasion resistance. The smooth interior contact surface wicks moisture away from the body for added comfort. The highly-efficient filtration media also provides greater air permeability of 8.65 cfm at 20 Pa, as tested in accordance with ASTM D737.