Interscience Laboratories 132-225 Baglight Multilayer Paddle Blender Bags Without Filter, 400 mL

Representative image only.
Interscience Laboratories Mfr # 132-225 – Item # EW-36850-34
  • Ultra-resistant multilayer®, multicoated complex
  • Rigid and transparent
  • Reinforced weldings

SPECS & DESCRIPTION

Specifications & Description

  • Bag TypePaddle Blender Bags
  • Bag Capacity (mL)400
  • Bag Capacity (L)0.4
  • DescriptionBaglight Multilayer Paddle Blender Bags Without Filter, 400 mL
Conformite EuropeenneISO9001:2000 Certified Supplier

Key Features

  • Ultra-resistant multilayer®, multicoated complex
  • Rigid and transparent
  • Reinforced weldings

More About this Item

Choose from a variety of bags to use with your paddle blender. Blending bags are available in various sizes and come with and without filters.

For any kind of sample preparation not requiring filtration.

PART OF A COMPLETE SYSTEM – GET EVERYTHING YOU NEED
The BagMixer® is the most effective lab blender for microbiological analysis. It guarantees optimal bacterial extraction of solid samples. Thanks to the unique adjustable paddle system, the blending chamber adapts itself perfectly to the nature and the size of the sample and works for any application.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat