Ismatec CA Pressure Lever Cassette Cartridge, 2-Stop, PVDF
Representative image only.
Ismatec Item # EW-78001-96
Choose new cassettes based upon material of construction, tubing type and pressure capabilities
  • Material of construction standard POM-C or optional PVDF
PRODUCT OPTIONS
$112.00USD / Each
BUY MORE & SAVE
  • 1-7$112.00USD / Each
  • 8-15$100.80USD / Each
  • 16-23$95.20USD / Each
  • 24+$89.60USD / Each
$154.00USD / Each
for an additional
$154.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Series2-Stop
  • Tubing sizes accepted0.13 to 3.19 mm ID
  • Max Pressure (bar)1.5
  • Max Pressure (PSI)21.7
  • Housing MaterialPOM-C
  • DescriptionPressure Lever Cassette / Cartridge, 2-Stop, PVDF
Key Features
  • Material of construction standard POM-C or optional PVDF
More About this Item
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.