Ismatec CA Tubing Retaining Adapters, 2-Stop, POM-C; 4/Pk
Representative image only.
Ismatec Item # EW-07618-67
Choose new cassettes based upon material of construction, tubing type and pressure capabilities
  • Material of construction standard POM-C or optional PVDF
PRODUCT OPTIONS
$40.00USD / Pkg of 4
BUY MORE & SAVE
  • 1-24$40.00USD / Pkg of 4
  • 25-99$36.00USD / Pkg of 4
  • 100+$34.00USD / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • Series2-Stop
  • Tubing sizes accepted0.13 to 3.19 mm ID
  • Housing MaterialPOM-C
  • DescriptionTubing Retaining Adapters, 2-Stop, POM-C, 4/pk
Key Features
  • Material of construction standard POM-C or optional PVDF
More About this Item
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.