This item is now discontinued. Please browse similar items
Ismatec Foot Switch for the Flowmaster Series of Pumps from
Representative image only.
Ismatec Item # EW-78016-97
Stainless steel construction—ideal for sanitary fluid transfer
  • Housing is IP65 rated
  • Change tubing in just seconds!
  • Easy to disassemble for cleaning
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFoot Switch for the Flowmaster Series of Pumps from
Includes
6-ft (1.8-m) cord with plug
Key Features
  • Housing is IP65 rated
  • Change tubing in just seconds!
  • Easy to disassemble for cleaning
More About this Item
This pump meets the strict hygienic requirements of the biotech, food and beverage, and pharmaceutical industries. The three-roller pump head with interchangeable rotors handles tubing with inside diameters of 6.4 to 16.0 mm and wall thicknesses of 1.6 and 3.2 mm. Accepts a 0 to 5 VDC, 0 to 10 VDC, 1 to 20 mA, or 4 to 20 mA input signal for remote speed control. Features multiple overload protection and an extensive analog interface with status information for process control systems.