Ismatec IS3510 Click-N-Go Cassette/Cartridge, 3-Stop, POM-C
Representative image only.
Ismatec Item # EW-78016-98
Choose new cassettes based upon material of construction, tubing type and pressure capabilities
  • Material of construction standard POM-C or optional PVDF
PRODUCT OPTIONS
$64.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Series3-Stop
  • Tubing sizes accepted0.13 to 3.19 mm ID
  • Max Pressure (PSI)14.5
  • Max Pressure (bar)1
  • Housing MaterialPOM-C
  • DescriptionClick-n-go Cassette / Cartridge, 3-Stop, POM-C
Key Features
  • Material of construction standard POM-C or optional PVDF
More About this Item
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.