Ismatec Miniflex Pump with Hand-Held Remote, 0.1 to 350 rpm; 90 to 260 VAC
Representative image only.
Ismatec Item # EW-77922-22
Complete systems to increase the efficiency and accuracy of filling and dispensing applications
  • Flow range 0.16 to 365 mL/min with included L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Realize increased efficiency and throughput when compared to hand pouring, manually pipetting, or using bottletop style dispensers
  • 5” (12.7 cm) capacitive TFT/RGB touch screen puts the intuitive interface at your fingertips
  • Dispense by volume or time -- anti-drip and speed ramping available in Volume dispense mode
  • Flexible operational control via screen, MasterflexLive, or analog remote control
PRODUCT OPTIONS
$3,190.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeVariable-Speed Digital
  • Min RPM0.1
  • Max RPM350
  • Number of Channels1
  • Max Flow Rate (mL/min)365
  • Min Flow Rate (mL/min)0.16
  • Max Flow Rate Per Channel (mL/min)365
  • Min Flow Rate Per Channel (mL/min)0.16
  • Drive78018-40
  • Pump HeadMiniflex
  • Tubing96440-25, 25-ft (BioPharm Plus, L/S® 25)
  • Tubing sizes acceptedL/S® 13, 14, 16, 25
  • Included AccessoriesHandheld remote controller and PTFE tube weight
  • Drive IP ratingIP 31
  • Washdown CapableNo
  • Power (VAC)90 to 260
  • Power (Hz)50/60
  • Open-Head SensorYes
  • Housing MaterialABS Plastic and Aluminum
  • Remote Inputs0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
Two year warranty
Includes
digital pump drive with Miniflex® pump head, handheld remote controller, PTFE tube weight, and 7.6 m (25 ft) of Masterflex L/S 25 BioPharm Plus silicone tubing. Also includes dual-voltage (115/230 VAC) benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination.
Key Features
  • Flow range 0.16 to 365 mL/min with included L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Realize increased efficiency and throughput when compared to hand pouring, manually pipetting, or using bottletop style dispensers
  • 5” (12.7 cm) capacitive TFT/RGB touch screen puts the intuitive interface at your fingertips
  • Dispense by volume or time -- anti-drip and speed ramping available in Volume dispense mode
  • Flexible operational control via screen, MasterflexLive, or analog remote control
  • Pump lets you save and retrieve commonly used dispense protocols for rapid set up
More About this Item
We’ve combined our Reglo Miniflex dispensing pump with everything you need to deliver accurate, repeat volumes for dispensing and filling applications. Two complete systems are available—with hands-free foot switch or handheld remote controller.
These systems also include Masterflex L/S 25 BioPharm Plus pump tubing. BioPharm Plus tubing offers low spallation, long pumping life, and exceptional flow stability over time—making it ideal for accurate filling and dispensing applications. Tubing is supplied in three eight-foot (2.4 m) lengths.