J. H. Williams Tool Group 1501MRMH Torque Wrench, 1/4in Drive, 20/150in.LB Micro-adj. comfort grip
Representative image only.
J. H. Williams Tool Group Mfr # 1501MRMH – Item # EW-97610-04
  • FEATURES
    • Dual scale, calibrated dual direction
    • Positive lock with spring loaded pull down lock ring
    • Fast, accurate and easy to set
    • Easy to read laser marked scales
    • Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
    • Accuracy: ± 4% clockwise ± 6% CCW of indicated value, CW, from 20 to 100% of full scale
    • Meets or exceeds ASME B107.14M-1994, ISO 6789 standards
    • Individually serialized with matching certificate of calibration traceable to NIST
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (N-m)5.4
  • Resolution (lb-in)50
  • Drive (in)0.25
  • Length (in)10.15
  • Capacity50 in lb / 5.4 N m
  • DescriptionTorque Wrench, 1/4in Drive, 20/150in.LB Micro-adj. comfort grip
Key Features
  • FEATURES
    • Dual scale, calibrated dual direction
    • Positive lock with spring loaded pull down lock ring
    • Fast, accurate and easy to set
    • Easy to read laser marked scales
    • Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
    • Accuracy: ± 4% clockwise ± 6% CCW of indicated value, CW, from 20 to 100% of full scale
    • Meets or exceeds ASME B107.14M-1994, ISO 6789 standards
    • Individually serialized with matching certificate of calibration traceable to NIST
More About this Item
Micro-adjustable torque wrenches; metal handle English scale
The metal handle series offers the user a rugged, industrial strength torque wrench able to withstand the rigors of professional use. handles are lightly knurled for a tough, non-slip grip. There are 34 models of metal handle micrometer adjustable torque wrenches available