J. H. Williams Tool Group 2001-I-MT Transducer 20-200 inLb Multitorq Sensor
Representative image only.
J. H. Williams Tool Group Mfr # 2001-I-MT – Item # EW-59872-74
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)200
  • Accuracy±1% of reading
  • Data Storage3500 readings
  • Data OutputRS-232
  • Capacity200 lb in
  • DescriptionTransducer 20-200 inLb Multitorq Sensor
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This is a small compact and accurate electronic meter capable of acquiring data from a variety of torque sensors and logging values into memory. It has been designed to be used for torque verification in assembly and manufacturing.
The unit can be used for auditing and process reporting while giving statistical process control (SPC) of stored data.
It can can also be used with in-line transducers resulting in the accuracies associated with assembling with an electronic torque wrench.