J. H. Williams Tool Group 2401CI3 Torque Wrench, 1/4" drive, 2.0 to 20.0 foot pounds
Representative image only.
J. H. Williams Tool Group Mfr # 2401CI3 – Item # EW-10780-47
Ergonomically designed molded non-slip rubber grip
PRODUCT OPTIONS
$568.00USD / Each
$79.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (kg-mm)27.6
  • Capacity (N-m)27.1
  • Capacity (lb-ft)20
  • Accuracy 2% cw of readings from 20% to 100% of full scale, 3% ccw of readings from 20% to 100% of full scale
  • Display3-color led light bar
  • Weight (lbs)2
  • Length (in)15
  • DescriptionTorque Wrench, 1/4" drive, 2.0 to 20.0 foot pounds
More About this Item
The COMPUTORQ3 Electronic Torque Wrench is a simple to use digital readout wrench that displays real time torque values in any of four torque units, ft. lbs., in. lbs., Nm, kg. cm.
Simply set the desired torque value and apply force until the green LED illuminates. Great for light industrial, automotive, motorcycle, watercraft, and aircraft applications.
* Sleek new design
* 15 degree flex ratchet in head
* Push button socket release head
* Large, easy to read digital display
* Long battery life, uses two each CR123 lithium batteries
* Torques in both clockwise and counterclockwise directions
* Battery saving auto-shutoff after two minutes
* COMPUTORQ3 remembers last unit and torque setting selected
* 3 color LED light bar
* When desired torque value is reached, display will hold value and