J. H. Williams Tool Group 6002LDIN Dial Indicating Torque Wrench, 3/8" Drive, 0-600 in lb
Representative image only.
J. H. Williams Tool Group Mfr # 6002LDIN – Item # EW-97150-25
$306.50USD / Each
$55.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)600
  • Capacity (N-m)70
  • Drive (in)0.375
  • Weight (lbs)1.75
  • Length (in)15
  • DescriptionDial Indicating Torque Wrench, 3/8" Drive, 0-600 in lb
More About this Item
Memory Needle Dial Memory needle acts as an indicator of how much torque has been applied. The pointer needle returns to zero but the memory needle remains at the highest applied torque value. Electric Dial (Alarm) Operator can preset a desired torque value. When the pointer needle reaches the pre set torque value, the light is illuminated and a buzzer sounds.
FEATURES
  • Exclusive bi-polar dual scales, English and Metric, permits accurate reading of both scales
  • Equally accurate in both left and right hand direction
  • Torsion drive design
  • Large easy to read dial
  • User assured of long life and reliability
  • Calibration traceable to National Institute of Standards and Technology (NIST)
  • Available with Follower Needle, Electric Light and Beeper or Ratcheting Drive (LDFNR)
  • Special dual scale combinations, (torque ranges & configurations)
  • Includes plastic case