Jeio Tech Lab Companion AAA44522 Universal Platform For 25l Shaking Bath
Representative image only.
Jeio Tech Lab Companion Mfr # AAA44522 – Item # EW-12054-42
  • 2 year warranty
PRODUCT OPTIONS
$350.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUniversal Platform For 25l Shaking Bath
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Change your shaker's capacities with convenient parts and accessories.
Maximum flask capacities:
(12) 50mL flask clamps
(9) 100mL flask clamps
(4) 250mL flask clamps
(4) 500mL flask clamps
(2) 1000mL flask clamps
(1) 2000mL flask clamp