Jenway 050 501 Fluorimeter Dataway PC Software, CD-ROM
Representative image only.
Jenway Item # EW-83061-23
Optimize performance of your Jenway® spectrophotometer
  • Expand analysis capabilities or make testing easier
  • Suitable for Windows, 2000, ME and XP operating systems
  • Unlimited storage of results and graphs
  • Powerful graphing function and flexible printing options
PRODUCT OPTIONS
$243.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithJenway 62 Series Fluorimeters
  • Width (cm)20.5
  • Height (cm)16.5
  • Length (cm)8.5
  • Power (VAC)100 to 240
  • DescriptionFluorimeter Dataway PC Software, CD-ROM
Key Features
  • Expand analysis capabilities or make testing easier
  • Suitable for Windows, 2000, ME and XP operating systems
  • Unlimited storage of results and graphs
  • Powerful graphing function and flexible printing options
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
Dataway gives you unlimited storage of results and graphs on your PC. Export your data into Excel format , or copy and paste your data into other programs. Dataway has its own powerful graphing function that enables multiple overlays of comparative data as well as the live up-date of data when continuous sampling is enabled (from the PC or instrument). High and low alarm levels can be displayed with incident reports when the graph crosses these thresholds.
With its flexible print-out options, Dataway will often be the only tool you need to record and present your results from the 62 Series Fluorimeters.