Jenway 628-200 Fluorimeter Heated Cell System, 100-240 VAC
Representative image only.
Jenway Item # EW-83061-22
Optimize performance of your Jenway® spectrophotometer
  • Expand analysis capabilities or make testing easier
  • Maintain your sample integrity
  • Perfect for temperature sensitive assays
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithJenway 62 Series Fluorimeters
  • Width (cm)20.5
  • Height (cm)16.5
  • Length (cm)8.5
  • Power (VAC)100 to 240
  • DescriptionFluorimeter Heated Cell System, 100-240 VAC
Key Features
  • Expand analysis capabilities or make testing easier
  • Maintain your sample integrity
  • Perfect for temperature sensitive assays
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
The electrically heated cell holder with its digital control unit ensures samples are always at the same temperature, decreasing the variance. Control the rate of your enzymatic reactions and other experiments requiring superior minimization of variance.
The heated cell system comes with the heated cell holder, controller, and connection leads for universal power supplies.