Jenway 736-201 Cuvette Holder, Water Heated, 10 mm Square
Representative image only.
Jenway Item # EW-83070-51
Optimize performance of your Jenway® spectrophotometer
  • Expand analysis capabilities or make testing easier
PRODUCT OPTIONS
$780.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithJenway 7300, 7305, 7310, 7315, and Genova Plus Spectrophometers
  • DescriptionCuvette Holder, Water Heated, 10 mm Square
Key Features
  • Expand analysis capabilities or make testing easier
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
Theis single-position cell/sample holder is compatible with the 73 series, as well as the Genova Plus and Nano lines of spectrophotometers. The water inlet and outlet lines allow you to connect the holder to a water line for temperature control of your sensitive samples. The holder accepts 10 x 10 mm square cuvettes for a 10 mm pathlength.