John Guest Push-to-Connect Adapter, Polypropylene, 3/8" x 1/4" NPT(M); 10/PK
Representative image only.
John Guest Mfr # PP011222W – Item # EW-34007-44
No need to buy two fitting halves
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
PRODUCT OPTIONS
$33.75USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Seal MaterialEPDM
  • Sterilization MethodAutoclave only
  • Connection TypePush-to-Connect x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/4
  • Push-to-Connect Tube OD (port 1) (in)3/8
  • Min Temperature (° F)33
  • Max Temperature (° F)140
  • Min Temperature (° C)1
  • Max Temperature (° C)60
  • Max Operating Pressure (PSI)150 psi at 70°F, 60 psi at 140°F (for water)
  • DescriptionPush-to-connect male adapter, polypropylene, 3/8" x 1/4" NPTM, 10/pack
Key Features
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
More About this Item
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth— tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.