John Guest Push-To-Connect Fitting, Male Threaded Adapter, Polypropylene, 1/4" OD x 1/8" NPT(M); 10/Pk
Representative image only.
John Guest Mfr # PP050821W – Item # EW-34007-53
Quick, leakproof connection
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
PRODUCT OPTIONS
$11.10USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • O-Ring MaterialEPDM
  • Collet MaterialStainless Steel
  • Sterilization MethodAutoclave only
  • Connection TypePush-to-Connect x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/8
  • Push-to-Connect Tube OD (port 1) (in)1/4
  • Min Temperature (° F)33
  • Max Temperature (° F)140
  • Min Temperature (° C)1
  • Max Temperature (° C)60
  • Max Operating Pressure (PSI)150 psi at 70°F, 60 psi at 140°F for water
  • DescriptionPush-To-Connect Fitting, Male Threaded Adapter, Polypropylene, 1/4" OD x 1/8" NPT(M); 10/Pk
Key Features
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
More About this Item
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters let you permanently modify basic fittings for customized installations.