Julabo Tubing Insulation, 12 mm ID; 1 m

Representative image only.
Julabo Mfr # 8930412 – Item # EW-12147-51
Quality accessories for Julabo Recirculators and Presto systems
  • Use from -50 to 100 °C
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Bath TypeBath/Circulator Accessory
  • DescriptionTubing Insulation, 12 mm ID; 1 m
Two year warranty

Key Features

  • Use from -50 to 100 °C
  • 2 year warranty

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat