Justrite 7150100 Safety Can for Flammables, Type I, 5 gallon

Representative image only.
Justrite Mfr # 7150100 – Item # EW-81791-43
Safely contain hazardous liquids and control vapors
  • A necessary part of compliance for safety in the workplace
  • Reduce the risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations

PRODUCT OPTIONS

$78.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialSteel
  • Capacity (Gal)5
  • Capacity (Liters)18.93
  • Exterior Height (in)16.875
  • Exterior Diameter (in)11.75
  • Exterior Height (cm)42.86
  • Exterior Diameter (cm)29.85
  • ComplianceNFPA, OSHA
  • ApprovalFM, UL, cUL
  • DescriptionSafety Can for Flammables, Type I, 5 gallon

Key Features

  • A necessary part of compliance for safety in the workplace
  • Reduce the risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations

More About this Item

All cans include a stainless steel flame arrester that prevents flashback ignition to reduce fire risks; also include a large ID zone for user labeling of contents to avoid misuse. Self-closing caps are designed with a relief mechanism to vent pressure — protects against rupture and explosion.

Type I safety cans feature a single spout for filling and dispensing. Spring-mounted caps are designed with a relief mechanism to vent pressure when it reaches 5 psig—protects against dangerous pressure build-up. A 3-1/2" long stainless steel flame arrester prevents flashback ignition, reducing fire risk. Optional "stay-on" polypropylene funnel fits Type I steel cans.

Approval: FM, UL, cUL

Compliance: OSHA, NFPA

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat