Justrite 891200 Compact Cabinets with manual doors, 12 gallon capacity

Representative image only.
Justrite Mfr # 891200 – Item # EW-81790-23
Compact cabinets are ideal for benchtop use

PRODUCT OPTIONS

$761.25USD / Each
Often Bought with...
Cole-Parmer ThinTouch™ Nitrile Gloves
$9.00 - $79.50USD / Case of 1000

SPECS & DESCRIPTION

Specifications & Description

  • Material18-gauge steel
  • Capacity (Liters)45.4
  • Capacity (Gal)12
  • Number of Shelves1
  • Width (in)24 1/4
  • Height (in)35
  • Depth (in)18
  • Width (cm)61.6
  • Height (cm)88.9
  • Depth (cm)45.7
  • ColorYellow
  • Self-ClosingNo
  • ComplianceOSHA, NFPA
  • Shipping Weight (lbs)131
  • DescriptionCompact Cabinets with manual doors, 12 gallon capacity

More About this Item

Use these 18-gauge steel constructed cabinets to safely store flammable or combustible liquids. Cabinets come equipped with one adjustable shelf for convenience. Doors have a three point fail-safe locking mechanism and feature a “Flammable” hazard label. Place cabinets under an existing countertop or use as stand alone units.


Compliance: OSHA/NFPA, UFC 79

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat