DWK Life Sciences (Kimble) 20024-01 Graduated Glass Cylinder Starter Pack, to deliver
Representative image only.
DWK Life Sciences (Kimble) Mfr # 20024-01 – Item # EW-34547-20
$273.50USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • DescriptionGraduated Glass Cylinder Starter Pack, to deliver
More About this Item
Kimax® cylinder assortment pack are calibrated ''to deliver''. The pack consists of one each of the 10mL, 25mL, 50mL, 100mL, and 250mL sized cylinders. Bumper guards are included for the 25mL through 250mL cylinders.