Kimberly-Clark WypAll® X80 Wiper, 9.1" x 16.8", 80/Box, 5 Boxes/Case

Representative image only.
Kimberly-Clark Mfr # 10351437 – Item # EW-33670-35
Design to be durable, WypAll® Wipers withstand solvents
  • Heavy duty machine and part wiping
  • Wiping metal shavings
  • Prepping surfaces with solvents
  • Cleaning rough surfaces
  • Heavy duty machine and part wiping
  • Wiping metal shavings
  • Prepping surfaces with solvents
  • Cleaning rough surfaces

PRODUCT OPTIONS

$196.25USD / Case of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialSoft pulp fibers, bonded to a polypropylene base sheet
  • ColorBlue
  • Width (in)9 1/10
  • Length (in)16 51/64
  • Width (cm)23.114
  • Length (cm)42.672
  • DescriptionWypall Wiper, model X80, 9.1\" x 16.8\", 80/box, 5 boxes/case

Key Features

  • Heavy duty machine and part wiping
  • Wiping metal shavings
  • Prepping surfaces with solvents
  • Cleaning rough surfaces
  • Heavy duty machine and part wiping
  • Wiping metal shavings
  • Prepping surfaces with solvents
  • Cleaning rough surfaces

More About this Item

The ultimate heavy-duty wiper. Towels are softer and more absorbent than general shop towels. Ideal for heavy-duty machine wiping, cleaning rough surfaces, wiping metal shavings, and solvent cleaning.

The ultimate heavy-duty wiper. Towels are softer and more absorbent than general shop towels. Ideal for heavy-duty machine wiping, cleaning rough surfaces, wiping metal shavings, and solvent cleaning.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat