DWK Life Sciences (Kimble) KC45066-16125 Culture Tube, plastic-coated glass, 16 mL, 144/cs
Representative image only.
DWK Life Sciences (Kimble) Mfr # KC45066-16125 – Item # EW-34572-32
  • Class A glass gives uniform thickness
PRODUCT OPTIONS
$863.00USD / Case of 144
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes
  • Tube Capacity (mL)16
  • Tube MaterialGlass
  • Tube ColorClear
  • Tube Bottom ShapeRound
  • Tube Length (mm)125
  • Tube Outside Diameter (OD) (mm)16
  • Cap IncludedYes
  • Cap StyleScrew Cap
  • GraduationsNone
  • DescriptionCultureTube,plastic-coatedglass,16mL,144/cs
Key Features
  • Class A glass gives uniform thickness
More About this Item
Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids. Glassware has a unique nonslip surface when wet or dry.Manufactured from KG-33 borosilicate glass for maximum heat transfer and superior chemical resistance. Meets ASTM E982, Type VI requirements. Rubber-lined screw caps are included and are unattached. Autoclavable to120°C (250°F).