Kinesis Hollow Cathode Lamp, phosphorous, 50 mm, Perkin Elmer Lumina coded
Representative image only.
Kinesis Item # EW-11951-59
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
  • Carry out quicker start-up times Reduce your operating costs with longer lifetime amp hours Continuous light intensity for longer periods of time Data-coded and uncoded versions are available
PRODUCT OPTIONS
$738.00USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPhosphorous
  • Compatible WithPerkin Elmer Lumina
  • Description Hollow Cathode Lamp, phosphorous, 50 mm, Perkin Elmer Lumina coded
Key Features
  • Carry out quicker start-up times Reduce your operating costs with longer lifetime amp hours Continuous light intensity for longer periods of time Data-coded and uncoded versions are available
More About this Item
Kinesis phosphorous (P) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.