Kinesis KX+ LCGC Fused-Insert Screw Top Vials, Glass, 9 mm Short Thread, 0.3 mL, PTFE/Silicone Septum; 100/pk
Representative image only.
Kinesis Item # EW-98701-56
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Fused-insert vials
  • PTFE/silicone septum
PRODUCT OPTIONS
$183.25USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)0.3
  • Tube MaterialGlass
  • Tube ColorClear
  • Cap TypeShort Thread Screw
  • Cap Diameter (mm)9
  • Septum TypePTFE/silicone
  • SterileNo
  • Special FeaturesFused Insert
  • DescriptionLCGC Fused-Insert Screw Top Vials, Glass, 9 mm Short Thread, 0.3 mL, PTFE/Silicone Septum; 100/pk
Includes
Screw top caps with septa and certificate of analysis.
Key Features
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Fused-insert vials
  • PTFE/silicone septum
More About this Item
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Glass vials are made of Type I, Class A 33-expansion borosilicate glass. Fused-insert vials are ideal for handling small-volume samples while delivering low residual volume (<8 µL). Total-recovery vials deliver maximum sample capacity (1.2 mL) with minimal residual volume (<1 µL)—use with bottom or side draw-port needles.