Cole-Parmer KX+ LCGC Screw Top Vials, Glass, 9 mm Short Thread, 2 mL, with Label Area, Pre-Slit PTFE/Silicone Septum; 100/pk
Representative image only.
Cole-Parmer Item # EW-98701-93
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Vials are constructed of clear glass vials with label
  • Pre-slit PTFE/silicone septum
PRODUCT OPTIONS
$66.00USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)2
  • Tube MaterialGlass
  • Tube ColorClear
  • Cap TypeShort Thread Screw
  • Cap Diameter (mm)9
  • Septum TypePre-slit PTFE/silicone
  • SterileNo
  • Special FeaturesLabel Area
  • DescriptionLCGC Screw Top Vials, Glass, 9 mm Short Thread, 2 mL, with Label Area, Pre-Slit PTFE/Silicone Septum; 100/pk
Key Features
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Vials are constructed of clear glass vials with label
  • Pre-slit PTFE/silicone septum
More About this Item
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Clear glass vials are made of Type I, Class A 33-expansion borosilicate glass. Amber vials are made of Type I, Class B 51-expansion borosilicate glass and are ideal for light-sensitive samples.