Kinesis KX Membrane Filter, Nylon, 47 mm dia., 0.8 µm; 100/pk
Representative image only.
Kinesis Item # EW-12917-80
Remove particulate without adversely affecting your final product
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • Pore Size (µm)0.8
  • Diameter (mm)47
  • DescriptionMembrane Filter, Nylon, 47 mm dia., 0.8 µm; 100/pk
Key Features
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results