Kinesis PTFE/Silicone Septum, for 15-425 thread caps; 100/pk
Representative image only.
Cole-Parmer Item # EW-98706-03
Choose the right septum for optimal performance and results
  • Various materials, thicknesses and hardnessess to suit your application
  • Pre-slit septa prevent a vacuum forming inside the vial
  • Natural PTFE/clear silicone, fits 15-425 thread caps
PRODUCT OPTIONS
$61.50USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Septum TypeNatural PTFE/clear silicone
  • Special Features2.6 mm thick
  • DescriptionPTFE/Silicone Septa, 15 mm; 100/pk
Key Features
  • Various materials, thicknesses and hardnessess to suit your application
  • Pre-slit septa prevent a vacuum forming inside the vial
  • Natural PTFE/clear silicone, fits 15-425 thread caps
More About this Item
Red rubber/Polytetrafluoroethylene (PTFE): improved level of cleanliness compared to natural rubber. PTFE coating ensures compatibility.
Natural rubber/Ethylene Tetrafluoroethylene (TEF): features excellent resealing properties.
Silicone/PTFE: cleanest solution for contaminant free sample handling; autoclavable, too.
PTFE/Silicone/PTFE: PTFE laminated to both sides of high-purity silicone to prevent coring.