Cole-Parmer Red Aluminum Crimp Cap, 11 mm Dia., Rubber/Butyl/TEF Septum w/Hole; 1000/pk
Representative image only.
Cole-Parmer Item # EW-98704-43
Aluminum crimps tightly around the neck for a tight seal
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Red aluminum 11 mm dia. cap Natural rubber/butyl/PTFE septum with a center hole Septum Hardness: 45° shore A Thickness: 1.0 mm
PRODUCT OPTIONS
$116.00USD / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeCrimp
  • Cap Diameter (mm)11
  • Cap MaterialAluminum
  • Cap colorRed
  • Septum TypeRubber/Butyl/PTFE
  • SterileNo
  • DescriptionCrimp Cap, 11mm, Red Aluminum, Rubber/Butyl/PTFE Septa; 1000/pk
Key Features
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Red aluminum 11 mm dia. cap Natural rubber/butyl/PTFE septum with a center hole Septum Hardness: 45° shore A Thickness: 1.0 mm
More About this Item
Constructed of aluminum, these crimp vials are ideal for use with your crimp neck vials. To ensure autosamplers function reliably, these caps and septa meet the rigorous specifications demanded by both the analysis and the instrument itself. Defined hardness and cleanliness properties ensure there is a septum available to be both fully compatible with the autosampler needle and meet the demands of the specific analysis.