Kinesis Short Thread Cap, 9mm Blue Polypropylene, Pre-Slit Silicone/Red PTFE Septa; 1000/pk SKU 9870312
Representative image only.
Kinesis Item # EW-98703-12
No more damaged autosampler needles or sample loss
  • Ensure septa is not deformed due to overtightening—extends the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond® septum to avoid sample contamination
  • Silicone/PTFE septa offers the cleanest solution for contaminant free sample handling
  • UltraBond septa is bonded without adhesive, avoiding contamination of the sample
  • Designed with a blue open cap with silicone and red slit PTFE septa
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeShort Thread Screw
  • Cap MaterialPolypropylene
  • Cap colorBlue
  • Septum TypeUltraBond Pre-Slit Silicone/Red PTFE
  • SterileNo
  • Description Short Thread Cap, 9mm, Open Blue Polypropylene, UltraBond Pre-Slit Silicone/Red PTFE Septa; 1000/pk
Key Features
  • Ensure septa is not deformed due to overtightening—extends the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond® septum to avoid sample contamination
  • Silicone/PTFE septa offers the cleanest solution for contaminant free sample handling
  • UltraBond septa is bonded without adhesive, avoiding contamination of the sample
  • Designed with a blue open cap with silicone and red slit PTFE septa
More About this Item
Short thread polypropylene caps fit short thread vials. Caps are available in different colors, septum materials, and open or closed configurations to suit your application. UltraBond cap and septum form a one-piece unit using a bonding process without adhesives for contamination-free results. Pre-slit septa prevent vacuum forming inside the vial.