Kinesis Silver Aluminum Cap, 11 mm Dia., Rubber/PTFE Septum; 1000/pk
Representative image only.
Kinesis Item # EW-98704-36
Aluminum crimps tightly around the neck for a tight seal
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Silver aluminum 11 mm dia. cap Red rubber/PTFE beige septum Septum Hardness: 45° shore A Thickness: 1.0 mm
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeCrimp
  • Cap Diameter (mm)11
  • Cap MaterialAluminum
  • Cap colorSilver
  • Septum TypeRed Rubber/Beige PTFE
  • SterileNo
  • DescriptionCrimp Cap, 11mm, Aluminum, Red Rubber/Beige PTFE Septa; 1000/pk
Key Features
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Silver aluminum 11 mm dia. cap Red rubber/PTFE beige septum Septum Hardness: 45° shore A Thickness: 1.0 mm
More About this Item
Constructed of aluminum, these crimp vials are ideal for use with your crimp neck vials. To ensure autosamplers function reliably, these caps and septa meet the rigorous specifications demanded by both the analysis and the instrument itself. Defined hardness and cleanliness properties ensure there is a septum available to be both fully compatible with the autosampler needle and meet the demands of the specific analysis.