Cole-Parmer TELOS® 96-Well Filtration Microplate™, 20 µm PE, Single Fritted, Populated
Representative image only.
Cole-Parmer Item # EW-06479-91
Designed for routine filtration and sample preparation
  • Individual well is designed to ensure elution volumes are kept to a minimum
  • Includes 20 µm polyethylene frits
  • Use for gravity or vacuum application
  • Wells are already populated—ideal for high throughput assays
PRODUCT OPTIONS
$105.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (µL)600
  • DescriptionTELOS® 96-Well Filtration Microplate™, 20 µm PE, Single Fritted, Populated
Key Features
  • Individual well is designed to ensure elution volumes are kept to a minimum
  • Includes 20 µm polyethylene frits
  • Use for gravity or vacuum application
  • Wells are already populated—ideal for high throughput assays
More About this Item
Choose from loose well or populated 96-well plates to suit your application needs.
Populated plates come with 96 wells already in place yet allow the flexibility to remove any unwanted wells.