Kinesis TELOS® Bulk Nonpolar SPE Media, flash octadecyl (C18); 100 g
Representative image only.
Kinesis Item # EW-06478-53
Save money by purchasing high quality sorbents in bulk quantities
  • Choose from a wide range of SPE media for packing your own columns
SPECS & DESCRIPTION
Specifications & Description
  • Media Mass (g)100
  • End CapNo
  • SorbentC18 (flash octadecyl)
Key Features
  • Choose from a wide range of SPE media for packing your own columns
More About this Item
Pack your column with a single sorbent or create your own multilayer column using bulk media. Sorbents come in 100 g or 1 kg bottles.