Kinesis TELOS Column Adapters for 1-, 3- and 6 mL Columns

Representative image only.
Kinesis Item # EW-06479-94
Optimize performance of your TELOS chromatography columns

PRODUCT OPTIONS

$36.80USD / Pkg of 10

SPECS & DESCRIPTION

Specifications & Description

  • ColorClear
  • Compatible With1 mL, 3 mL, or 6 mL columns
  • ConnectionsLuer
  • DescriptionTELOS Column Adapters for 1-, 3- and 6 mL Columns

More About this Item

Choose from a wide range of accessories for flash chromatography dry loading, filtration, and large volume processing.
Provide a secure seal for both the inlet and outlet of TELOS flash columns with the luer cap and plug. TELOS column luer cap can be used in conjunction with the appropriate size column for either room temperature reactions or short-term compound storage.
Easily and quickly adapt 1-, 3-, or 6-mL TELOS SPE columns the column adapter.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat