Cole-Parmer TELOS® neo™ PRP Fixed-Well Plate, 10 mg sorbent; 1/ea
Representative image only.
Cole-Parmer Item # EW-06474-57
Modular design lets you simply remove any unwanted wells and add sealing strips
  • Quickly obtain clean extracts and reduce time spent on method development
  • Faster method development saves you money
  • Water-wettable polymer is not affected by drying out
  • Applications: Nonpolar sorbent for acidic, neutral, and basic analyte extraction
PRODUCT OPTIONS
$273.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)10
  • Column Particle Size (µm)60
  • Bottom TypePolymer based
  • ApplicationsNonpolar
  • SorbentPRP
  • DescriptionPRP Fixed-Well Plate, 10 mg sorbent; 1/ea
Key Features
  • Quickly obtain clean extracts and reduce time spent on method development
  • Faster method development saves you money
  • Water-wettable polymer is not affected by drying out
  • Applications: Nonpolar sorbent for acidic, neutral, and basic analyte extraction
More About this Item
Cole-Parmer® TELOS® neo™ populated plates come with 96 wells already in place—ideal for high throughput assays. Polymer-based sorbents have no silanol interactions, excellent flow characteristics, and pH stability providing highly reproducible results. Elution volumes are kept to a minimum by allowing maximum interaction between the analyte and sorbent.