Cole-Parmer TELOS® Polyethylene Frits 10 µm, 70 mL

Representative image only.
Cole-Parmer Item # EW-06479-17
Optimize performance of your TELOS reservoirs
  • Available in 10 or 20 µm
  • 10 µm PE for 70 mL columns

PRODUCT OPTIONS

$142.00USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • Wetted MaterialsPolyethylene
  • Fritted Disk10 µm
  • DescriptionTELOS Polyethylene Frits 10 µm, 70 mL

Key Features

  • Available in 10 or 20 µm
  • 10 µm PE for 70 mL columns

More About this Item

Choose from multiple frit material options ideal for your application needs.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat