Cole-Parmer TELOS® Replacement Vacuum Gauge, Valve, and Glass Tank for Sample Processing Manifold, 12 position
Representative image only.
Cole-Parmer Item # EW-06479-80
Use manifold accessories to process multiple columns at once
  • Optimize performance of your sample processing manifolds
  • Easily control the flow rates by adjusting the vacuum supply to the manifold
$225.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithManifold 06480-04
  • DescriptionTELOS Replacement Vacuum Gauge, Valve, and Glass Tank for Sample Processing Manifold, 12 position
Key Features
  • Optimize performance of your sample processing manifolds
  • Easily control the flow rates by adjusting the vacuum supply to the manifold
More About this Item
Choose from a wide range of accessories including tanks, fittings, and more for your sample processing manifolds.