Cole-Parmer TELOS® Specialty SPE Column, Diol-Functionalized Silica, 200 mg, 3 mL; 50/Pk
Representative image only.
Cole-Parmer Item # EW-06476-16
Easily extract very polar analytes from nonpolar nonaqueous sample matrices
  • Diol-functionalized silica is a polar bonded SPE sorbent
$134.50USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)200
  • Total Volume Capacity (mL)3
  • SorbentDiol-functionalized silica
Key Features
  • Diol-functionalized silica is a polar bonded SPE sorbent
More About this Item
Diol-functionalized silica is a polar bonded SPE sorbent, less polar compared with silica and suitable for the extraction of very polar analytes from nonpolar nonaqueous sample matrices. It can also be used as a nonpolar sorbent for the extraction of relatively nonpolar analytes from aqueous sample matrices. Columns are made of polypropylene and feature Luer-slip outlet.