Cole-Parmer TELOS® Specialty SPE Column, Graphitized Carbon, 250 mg Sorbent, 3 mL; 50/Pk
Representative image only.
Cole-Parmer Item # EW-06478-24
Ideal for polar water-soluble analyte extraction from aqueous sample
  • Manufactured with graphitized carbon sorbent
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)250
  • Total Volume Capacity (mL)3
  • SorbentGraphitized carbon
Key Features
  • Manufactured with graphitized carbon sorbent
More About this Item
Graphitized carbon sorbent is a great for polar water-soluble analyte extraction from aqueous samples. It has an extremely high affinity for a very wide range of analytes using a nonpolar retention mechanism. Columns are made of polypropylene and feature Luer-slip outlet.