Cole-Parmer TELOS® Specialty SPE Column, PS-DVB (ENV), 200 mg Sorbent, 6 mL; 30/Pk
Representative image only.
Cole-Parmer Item # EW-06477-93
Ideal for polar water-soluble analyte extraction from aqueous samples
  • Manufactured with PS-DVB (polystyrene divinylbenzene resin) and PS-DVB ENV (hydroxylated polystyrene divinylbenzene resin) solvents
PRODUCT OPTIONS
$197.00USD / Pkg of 30
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)200
  • Total Volume Capacity (mL)6
  • SorbentPS-DVB (ENV)
Key Features
  • Manufactured with PS-DVB (polystyrene divinylbenzene resin) and PS-DVB ENV (hydroxylated polystyrene divinylbenzene resin) solvents
More About this Item
PS-DVB (polystyrene divinylbenzene resin) and PS-DVB ENV (hydroxylated polystyrene divinylbenzene resin) are ideal for polar water-soluble analyte extraction from aqueous samples. PS-DVB ENV has a very high surface area, offering excellent retention of very polar analytes including phenols and phenoxyacid herbicides from environmental samples and drug metabolites from biological fluids. Columns are made of polypropylene and feature Luer-slip outlet.