Cole-Parmer TELOS® Top Frit for Dry Loading Cartridge, Slip Tip, 80 g, 20 µm; 50/pk

Representative image only.
Cole-Parmer Item # EW-06475-41
Extend the life of your dry loading cartridges with these replacement parts
  • Constructed of durable polyethylene
  • Compatible with TELOS 80 g dry loading cartridge with slip tip
  • 20 µ fritted disk

PRODUCT OPTIONS

$253.00USD / Pkg of 50

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolyethylene
  • Compatible WithColumn 06475-63; 20 µm
  • DescriptionTop Frit for Dry Loading Cartridge, Slip Tip, 80 g, 20 µm; 50/pk

Key Features

  • Constructed of durable polyethylene
  • Compatible with TELOS 80 g dry loading cartridge with slip tip
  • 20 µ fritted disk

More About this Item

Choose from a variety of replacement top frit sizes for your Cole-Parmer TELOS dry loading columns.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat