Kinesis White Polypropylene EPA Screw Cap, 24mm, Closed Silicone/PTFE bonded Septa; 1000/pk
Representative image only.
Kinesis Item # EW-98700-43
Caps come preassembled with silicone/PTFE septa permanently bonded to the cap to reduce the chance of contamination
  • Select from open (injection), closed (storage), or SURE-Clean™ caps, depending on your application
  • Closed cap with silicone/PTFE bonded septum
PRODUCT OPTIONS
$608.00USD / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeScrew cap
  • Cap Diameter (mm)24
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Septum TypeBonded silicone/PTFE bonded
  • DescriptionWhite Polypropylene EPA Screw Cap, 24mm, Closed Silicone/PTFE bonded Septa; 1000/pk
Key Features
  • Select from open (injection), closed (storage), or SURE-Clean™ caps, depending on your application
  • Closed cap with silicone/PTFE bonded septum
More About this Item
All caps come with silicone/PTFE bonded septa. Each septum is 3.2 mm thick and had hardness rating of 45° shore A.