KleenGuard 44314 A40 Coveralls, Size X-Large, Case/25
Representative image only.
KleenGuard Mfr # 44314 – Item # EW-86414-12
Designed for comfort and extended wear, and in multiple styles to best fit your application
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • SizeX-Large
  • MaterialPolypropylene(PP)
  • ColorWhite
  • ComplianceNFPA 99
  • DescriptionA40 Coveralls, Size X-Large, Case/25
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
KleenGuard coveralls protect you from dry particulates and light liquid sprays. Three-layer fabric includes a layer of microporous film so heat and sweat vapors escape, allowing workers to stay with the task at hand with less fatigue and heat stress!
The concealed front zipper measures at least 28" long. Three-thread overlap stitching improves strength and durability. Coveralls can be easily donned or removed without tearing. Comfortable and generously cut coveralls are 7-1⁄2 times less likely to rip out than those made using the typical ANSI-standard design. Styles with elastic cuffs feature an elastic back waistband for improved fit and comfort.
A40 Coveralls: Breathable microporous film laminate guards worker from particulates and liquid, and is an excelle
Call our Application Specialists to receive a copy of our Chemical Resistance Charts.