KleenGuard 97824 Gray Nitrile Gloves, X-Large, Box/150
Representative image only.
KleenGuard Mfr # 97824 – Item # EW-86484-07
Get 50% more gloves per case, reduces packaging waste
PRODUCT OPTIONS
$68.40USD / Box of 150
BUY MORE & SAVE
  • 1-9$68.40USD / Box of 150
  • 10+$61.56USD / Box of 150
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)3.5
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGray
  • GripTextured
  • CuffRolled
  • SterileNo
  • DescriptionGray Nitrile Gloves, X-Large, Box/150
More About this Item
KleenGuard G10 grey nitrile gloves are latex-free and designed for tasks requiring maximum dexterity and tactile sensitivity. These gloves are also environmentally friendly since you get 50% more gloves per case — resulting in less packaging and added space savings. Gloves are powder-free, nonsterile, and have rolled cuffs. Finger tips are textured for added grip.